Learn More
Sino Biological Anti-IL-36 alpha/IL-1F6 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10607MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1F6 / IL36A (rh IL1F6 / IL36A; Catalog#10607-HNAE; Q9UHA7; Met 1-Phe 158). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8832