Learn More
Sino Biological Anti-IL-36 alpha/IL-1F6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10607T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-1F6 (rh IL-1F6; Catalog#10607-HNAE; Q9UHA7; Met1-Phe158). IL-1F6 specific IgG was purified by Human IL-1F6 affinity chromatography.
Catalog No. 50-241-1446