Learn More
Sino Biological Anti-IL-36 alpha/IL-1F6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10607RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL1F6 / IL36A (rh IL1F6 / IL36A; Catalog#10607-HNAE; Q9UHA7; Met 1-Phe 158). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1445