Learn More
Sino Biological Human IL36B PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10579T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL36B / IL1F8 (rh IL36B / IL1F8; Catalog#10579-HNAE; Q9NZH7-2; Met1-Glu157). IL36B / IL1F8 specific IgG was purified by Human IL36B / IL1F8 affinity chromatography.
Catalog No. 50-237-0845