Learn More
Sino Biological Recombinant Anti-IL-3R alpha/CD123 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10518R01220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL3RA / CD123 (rh L3RA; Catalog#10518-H08H; NP_002174.1; Met 1-Arg 305).
Catalog No. 50-241-1481