Learn More
BIOHIPPO Human IL3Ra Microsample
SDP
Catalog No. 5000313839
Encompass
Human IL3Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL3Ra in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 134 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL3Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL3Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL3Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13839 Supplier BIOHIPPO Supplier No. BHE15211595
Human IL3Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL3Ra in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 134 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL3Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL3Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL3Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color