Learn More
Sino Biological Human IL3Ra PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10518T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-3R alpha/CD123 (rh IL-3R alpha/CD123; Catalog#10518-H08H; NP_002174.1; Met1-Arg305). IL-3R alpha/CD123 specific IgG was purified by Human IL-3R alpha/CD123 affinity chromatography.
Catalog No. 50-237-0973