Learn More
Sino Biological Human IL3Ra Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10518H41HB100
Encompass_Preferred
A DNA sequence encoding the human IL3Ra (NP_002174.1)(Met1-Arg305) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2091