Learn More
Sino Biological Human IL4 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502371760
Encompass_Preferred
Each vial contains 7 ng of recombinant Human IL4 / IL-4. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 500 pg/mL is recommended.
Catalog No. 50-237-1760 Supplier Sino Biological Supplier No. SEKA118465
Each vial contains 7 ng of recombinant Human IL4 / IL-4. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 500 pg/mL is recommended.