Learn More
Sino Biological Recombinant Human IL-4 Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 11846HNAEB20
Encompass_Preferred
A DNA sequence encoding the Human IL4 (P05112-1) (His25-Ser153) was expressed with a N-terminal Met. The purified protein was biotinylated in vitro.
Catalog No. 50-224-6617