Learn More
Sino Biological Human IL4 Protein 20ug (Sino Biological)
Supplier:  Sino Biological GMP11846HNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of human IL4 isoform 1 (P05112-1) (His25-Ser153) was expressed and purified with an initial Met.
Catalog No. 50-237-3719