Learn More
Sino Biological Recombinant Human IL-4R Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10402H08HB100
Encompass_Preferred
A DNA sequence encoding the Human IL4R (NP_000409.1) (Met1-His232) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5316