Learn More
Sino Biological Recombinant Anti-IL-5 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 15673R225100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-5 (rh IL-5; Catalog#15673-HNCE; P05113; Ile20-Ser134).
Catalog No. 50-241-2231