Learn More
BIOHIPPO Human IL5Ra Microsample
SDP
Catalog No. 5000315043
Encompass
Human IL5Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL5Ra in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 24 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL5Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL5Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL5Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15043 Supplier BIOHIPPO Supplier No. BHE15211578
Human IL5Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL5Ra in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 24 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL5Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL5Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL5Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color