Learn More
Sino Biological Recombinant Human IL-5R alpha Protein (His Tag)
Supplier:  Sino Biological 10392H08H1
Encompass_Preferred
A DNA sequence encoding the human IL5Ra isoform 2 (NP_783852.1) (Met 1-Arg 335) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5330