Learn More
Sino Biological Human IL6 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370841
Encompass_Preferred
Each vial contains 44 ng of recombinant Human IL-6 / Interleukin-6. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 200 pg/mL is recommended.
Catalog No. 50-237-0841 Supplier Sino Biological Supplier No. SEKB1039515
Each vial contains 44 ng of recombinant Human IL-6 / Interleukin-6. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 200 pg/mL is recommended.