Learn More
Sino Biological Human IL6 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10395MM1420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-6 / Interleukin-6 (rh IL-6 / Interleukin-6; Catalog#10395-HNAE; NP_000591.1; Val30-Met212). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1987