Learn More
Sino Biological IL6 / IL-6 Neutralizing Antibody
Supplier:  Sino Biological 10395MHK23500
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant Human IL6 / IL-6. The antibody was produced using recombinant antibody technology.
Catalog No. 50-241-0097