Learn More
Sino Biological Anti-IL-6 Antibody (Biotin), Rabbit Polyclonal
Supplier:  Sino Biological 10395T16B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL6 / IL-6 (rh IL6 / IL-6; Catalog#10395-HNAE; NP_000591.1; Val30-Met212). IL6 / IL-6 specific IgG was purified by Human IL6 / IL-6 affinity chromatography and then biotinylated.
Catalog No. 50-241-1472