Learn More
Sino Biological Anti-IL-6R Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10398MM18A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-6R / CD126 (rh IL-6R / CD126; Catalog#10398-H08H; NP_000556.1; Met1-Pro365). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-5323