Learn More
Sino Biological Recombinant Anti-IL-6R Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10398R502A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-6R (rh IL-6R; Catalog#10398-H08H; NP_000556.1; Met1-Pro365), and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-5321