Learn More
Sino Biological Human IL6ST MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10974R014100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL6ST/gp130/CD130 (rh IL6ST/gp130/CD130; Catalog#10974-HCCH; NP_002175.2; Met1-Ile618).
Catalog No. 50-237-1367