Learn More
Sino Biological Anti-gp130/IL6ST Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10974RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human gp130 / CD130 / IL6ST (rh gp130 / CD130 / IL6ST; Catalog#10974-HCCH; NP_002175.2; Met1-Ile618). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5007