Learn More
Sino Biological Human IL6ST Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10974H05H100
Encompass_Preferred
A DNA sequence encoding the human IL6ST (AAA59155.1) (Met1-Ile618) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-237-1873