Learn More
Sino Biological Recombinant Anti-IL-7 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11821R022100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL7 / IL-7 / Interleukin-7 (rh IL7 / IL-7 / Interleukin-7; Catalog#11821-HNAE; NP_000871.1; Asp26-His177).
Catalog No. 50-240-7969