Learn More
Sino Biological Recombinant Human IL-7R alpha/CD127 Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10975H49HB20
Encompass_Preferred
A DNA sequence encoding the human IL7R (P16871-1) (Met1-Gly236, with mutations I66T, V138I) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme), which is co-expressed.
Catalog No. 50-228-2931