Learn More
Sino Biological Anti-IL-7R alpha/CD127 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10975MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL7RA (rh IL7RA; Catalog#10975-H08H; P16871-1; Met 1-Gly 236). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0347