Learn More
Sino Biological Recombinant Anti-IL-7R alpha/CD127 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10975R014A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL7R/IL-7R/CD127 (rh IL7R/IL-7R/CD127; Catalog#10975-H08H; P16871-1; Met1-Gly236) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-9744