Learn More
Sino Biological Recombinant Human IL-7R alpha/CD127 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10975H02H1
Encompass_Preferred
A DNA sequence encoding the human IL7R (P16871-1) (Met1-Gly236, with mutations I66T, V138I) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5005