Learn More
Sino Biological Recombinant Human IL-8/CXCL8 Protein (aa 23-99) 10 µg
Supplier:  Sino Biological 10098HNCH110
Encompass_Preferred
The 77 amino acid endothelial-cell derived form (Ala 23-Ser 99) of the mature human IL8 (NP_000575.1) was expressed and purified.
Catalog No. 50-224-6945