Learn More
Sino Biological Recombinant Human IL-8/CXCL8 Protein (aa 28-99) 1 mg
Supplier:  Sino Biological 10098HNCH21
Encompass_Preferred
The 72 amino acid residue form (Ser 28-Ser 99) of mature human IL8 (NP_000575.1) was expressed and purified.
Catalog No. 50-224-6929