Learn More
BIOHIPPO Human IL8Ra Microsample
SDP
Catalog No. 5000315933
Encompass
Human IL8Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL8Ra in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL8Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL8Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL8Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15933 Supplier BIOHIPPO Supplier No. BHE15212243
Human IL8Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL8Ra in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL8Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL8Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL8Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color