Learn More
Sino Biological Recombinant Human IL-9 Protein (His Tag)
Supplier:  Sino Biological 11844H08B1
Encompass_Preferred
A DNA sequence encoding the human IL9 (NP_000581.1) (Met1-Ile144) was expressed and purified, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4522