Learn More
Lonza Walkersville HIAEC – Human Iliac Artery Endothelial Cells
Supplier:  Lonza Walkersville CC2545
Encompass
Primary Human Iliac Artery (HIAEC) Endothelial Cells are isolated from normal tissue. Additional donor characteristics including e.g. heart disease, hypertension, or smoking history may be available. For further information please contact our Scientific Support Team. Clonetics Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system. Cryopreserved HIAEC are shipped in third passage.Cryopreserved HIAEC are guaranteed through 10 population doublings. HIAEC express CD31/105, von Williebrand Factor VIII, and are positive for acetylated low density lipoprotein uptake. All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Content:1 x Cryopreserved Ampule with ≥500,000 cells/vial
Catalog No. NC2581704