Learn More
Creative Biomart Anthrax immune globulin human
Catalog No. 502084052
Encompass_Preferred
The product is a human immune globulin derived from the plasma of humans immunized with BioThrax (adsorbed anthrax vaccine), which is then further purified.
Catalog No. 50-208-4052 Supplier Creative Biomart Supplier No. THP0001
The product is a human immune globulin derived from the plasma of humans immunized with BioThrax (adsorbed anthrax vaccine), which is then further purified.