Learn More
Raybiotech Inc Quantibody Human Immune Response Biomarker Array 1 Kit.
Supplier:  Raybiotech Inc QAHIMR11
Encompass_Preferred
Detects 40 Human Immune Response-related biomarkers. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: CD14, CD163, CD40 (TNFRSF5), CRP (C-Reactive Protein), E-Selectin, Fas (TNFRSF6/Apo-1), Fas Ligand (TNFSF6), GCSF, ICAM-1 (CD54), IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 R4 (ST2), IL-10, IL-12 p70, IL-13, IL-18, IL-2, IL-2 R alpha, IL-4, IL-6, IL-8 (CXCL8), Lipocalin-2 (NGAL), MCP-1 (CCL2), MCP-2 (CCL8), MIF, MIP-1 alpha (CCL3), MIP-1 beta (CCL4), Osteopontin (SPP1), PAI-1, Platelet Factor 4 (CXCL4)
Catalog No. 50-195-0839