Learn More
R&D Systems™ Human Immuno-Oncology Panel 1 Luminex™ Performance Premix Kit

Specifications
Specifications
| Quantity | 1 x 96-well Plate |
| Species | Human |
| Product Line | R&D Systems |
| Kit Contents | 1 x 96-well microplate and Magnetic antibody-coated beads |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.