Learn More
Sino Biological Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11233RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 gp120 (rv GP120; Catalog#11233-V08H;Ala29-Glu502; AFM77980.1). GP120 specific IgG was purified by HIV-1 gp120 affinity chromatography.
Catalog No. 50-224-7602