Learn More
Sino Biological Human Immunodeficiency Virus type 1 (HIV-1) Gag-p24 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11695MM13100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HIV-1 Gag-p24 (HIV-1 Gag-p24; Catalog#11695-V08E; ACI05538.1; Pro 133-Leu 363). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8249