Learn More
Sino Biological Human Immunodeficiency Virus type 1 (HIV-1) gp120 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11233MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HIV-1 gp120 (Catalog#11233-V08H; Ala29-Glu502; AFM77980.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8232