Learn More
Biomatik Corporation Human Immunoglobulin A2 (IgA2) CLIA Kit
Supplier:  Biomatik Corporation EKU09974
Encompass_Preferred
Human Immunoglobulin A2 (IgA2) CLIA Kit for detecting IgA2 in serum, plasma, saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.69-500ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01877. Target Synonyms: IGHA2; Immunoglobulin Heavy Constant Alpha 2; A2m Marker. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8390