Learn More
Biomatik Corporation Human Immunoglobulin Binding Protein 1 (IGBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU08362
Encompass_Preferred
Human Immunoglobulin Binding Protein 1 (IGBP1) ELISA Kit for detecting IGBP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P78318. Target Synonyms: IBP1; B-cell signal transduction molecule alpha 4; CD79a-binding protein 1; Protein phosphatase 2/4/6 regulatory subunit; Renal carcinoma antigen NY-REN-16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4527