Learn More
Bioworld Human Immunoglobulin G Lyophilized Powder >= 97%, 1 g
Supplier:  Bioworld 220700881
Encompass_Preferred
The gamma globulin preparations are supplied as a lyophilized powder. Purity levels as GG are analyzed by SDS-Page or Agarose electrophoresis.These preparations are especially useful as a co-precipitant in RIA methods employing polyethylene glycol or ammonium sulfate precipitation steps,or as a protein stabilizer,or as blocker in a number of immunoassays.These preparations are also useful as GG control solutions for biochemical analytical tests.Gamma globulin preparations reconstitute to pH 7.2 ± 0.2. Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-253-977