Learn More
Biomatik Corporation Human Immunoglobulin Lambda Like Polypeptide 5 (IglL5) ELISA Kit
Supplier:  Biomatik Corporation EKU09379
Encompass_Preferred
Human Immunoglobulin Lambda Like Polypeptide 5 (IglL5) ELISA Kit for detecting IglL5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B9A064. Gene ID: 100423062. Target Synonyms: Germline Immunoglobulin Lambda 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4863