Learn More
Sino Biological Anti-IMPA1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15998RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IMP1 / IMPA1 (rh IMPA1; Catalog#15998-H07E; P29218-1; Met1-Asp277). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2453