Learn More
Raybiotech Inc C-Series Human Inflammation Antibody Array 1 Kit.
Catalog No. 501948601
Encompass_Preferred
Detects 20 Human Inflammatory Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-3, IL-4, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, I-309 (TCA-3/CCL1), TIMP-2
Catalog No. 50-194-8601 Supplier Raybiotech Inc Supplier No. AAHINF14
Detects 20 Human Inflammatory Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), GCSF, GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-3, IL-4, IL-6, IL-7, IL-8 (CXCL8), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, I-309 (TCA-3/CCL1), TIMP-2