Learn More
Raybiotech Inc C-Series Human Inflammation Antibody Array 2 Kit.
Supplier:  Raybiotech Inc AAHINF28
Encompass_Preferred
Detects 20 Human Inflammatory Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: ICAM-1 (CD54), IL-6 R, IL-15, IL-16, IL-17A, IP-10 (CXCL10), MCP-1 (CCL2), MCP-2 (CCL8), M-CSF, MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-1 delta (CCL15), RANTES (CCL5), TGF beta 1, TNF alpha, TNF beta (TNFSF1B), TNF RI (TNFRSF1A), TNF RII (TNFRSF1B), PDGF-BB
Catalog No. 50-194-8605