Learn More
Raybiotech Inc G-Series Human Inflammation Antibody Array 3 Kit.
Supplier:  Raybiotech Inc AAHINFG34
Encompass_Preferred
Detects 40 Human Inflammatory Factors. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), GCSF, GM-CSF, ICAM-1 (CD54), IFN-gamma, I-309 (TCA-3/CCL1), IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-3, IL-4, IL-6, IL-6 R, IL-7, IL-8 (CXCL8), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-16, IL-17A, IP-10 (CXCL10), MCP-1 (CCL2), MCP-2 (CCL8), M-CSF, MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-1 delta (CCL15), RANTES (CCL5), TGF beta 1, TNF alpha, TNF beta (TNFSF1B), TNF RI (TNFRSF1A), TNF RII (TNFRSF1B), PDGF-BB, TIMP-2
Catalog No. 50-194-8609