Learn More
Raybiotech Inc G-Series Human Inflammation Array 3 Kit.
Supplier:  Raybiotech Inc GSHINF32
Encompass_Preferred
Semi-quantitative measurement of 40 human cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: BLC (CXCL13), Eotaxin-1 (CCL11), Eotaxin-2 (MPIF-2/CCL24), GCSF, GM-CSF, I-309 (TCA-3/CCL1), ICAM-1 (CD54), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 ra (IL-1 F3), IL-2, IL-4, IL-5, IL-6, IL-6 R, IL-7, IL-8 (CXCL8), IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-16, IL-17A, MCP-1 (CCL2), M-CSF, MIG (CXCL9), MIP-1 alpha (CCL3), MIP-1 beta (CCL4), MIP-1 delta (CCL15), PDGF-BB, RANTES (CCL5), TIMP-1, TIMP-2, TNF alpha, TNF beta (TNFSF1B), TNF RI (TNFRSF1A), TNF RII (TNFRSF1B)
Catalog No. 50-195-0411