Learn More
BIOHIPPO Human INHbB Microsample
SDP
Catalog No. 5000313844
Encompass
Human INHbB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of INHbB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human INHbB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human INHbB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human INHbB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13844 Supplier BIOHIPPO Supplier No. BHE15212098
Human INHbB Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of INHbB in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human INHbB Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human INHbB Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human INHbB biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color